DL-2,3-Diaminopropionic acid hydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
2,3-Diaminopropanoic acid hydrochloride, 3-Amino-DL-alanine Hydrochloride, DL-2,3-Diaminopropionic acid monohydrochloride
D0438-5G
D0438-1G
D0438-25G
TCD0438-005GEA
259.12
CAD
TCD0438-005G
TCD0438-001G
TCD0438-25G
DL-2,3-Diaminopropionic acid hydrochloride ≥98.0% (by total nitrogen basis)
DL-2,3-Diaminopropionic acid hydrochloride
CAS Number: 54897-59-5
MDL Number: MFCD00012884
Molecular Formula: C3H8N2O2
Molecular Weight: 140.57
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 237
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 3-Amino-DL-alanine Hydrochloride
Formula:
C₃H₈N₂O₂ MW: 140.57 g/mol Melting Pt: 237 °C |
MDL Number:
MFCD00012884 CAS Number: 54897-59-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...