Diphenhydramine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D0423-25G D0423-500G
TCD0423-025GEA 58.46 CAD
TCD0423-025G TCD0423-500G
Diphenhydramine hydrochloride ≥98.0% (by HPLC, titration analysis)
Diphenhydramine hydrochloride

CAS Number: 147-24-0
MDL Number: MFCD00012479
Molecular Formula: C17H21NO
Molecular Weight: 291.82
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 170


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents
Other Product Names / Synonyms: 2-Diphenylmethoxy-N,N-dimethylethylamine Hydrochloride, 2-(Benzhydryloxy)-N,N-dimethylethylamine Hydrochloride

Formula: C₁₇H₂₁NO
Melting Pt: 170 °C
MDL Number: MFCD00012479
CAS Number: 147-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dc7104d40b002a4abfc6896f635a636a5b28696&vid=0dc7104d40b002a4abfc6896f635a636a5b28696&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR