2',7'-Dichlorofluorescein ≥95.0%
Supplier: TCI America
Synonyms:
2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
D0371-25G
D0371-1G
TCD0371-025GEA
264.81
CAD
TCD0371-025G
TCD0371-001G
2',7'-Dichlorofluorescein ≥95.0%
2',7'-Dichlorofluorescein
[for Fluorescent indicator]
CAS Number: 76-54-0
MDL Number: MFCD00005047
Molecular Formula: C20H10Cl2O5
Molecular Weight: 401.20
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 280
Formula:
C₂₀H₁₀Cl₂O₅ MW: 401.20 g/mol Melting Pt: 280 °C |
MDL Number:
MFCD00005047 CAS Number: 76-54-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...