1,4-Dibenzyloxybenzene ≥98.0%

Supplier: TCI America

D0159-5G D0159-25G
TCD0159-5GEA 74.26 CAD
TCD0159-5G TCD0159-025G
1,4-Dibenzyloxybenzene ≥98.0%
1,4-Dibenzyloxybenzene

CAS Number: 621-91-0
MDL Number: MFCD00016875
Molecular Formula: C20H18O2
Molecular Weight: 290.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 128


Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 1,4-Bis(benzyloxy)benzene, Hydroquinone Dibenzyl Ether

Formula: C₂₀H₁₈O₂
MW: 290.36 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00016875
CAS Number: 621-91-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=235530498a6718421e82ed70ed1f4c3573d51770&vid=235530498a6718421e82ed70ed1f4c3573d51770&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR