2,6-Diaminopimelic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,5-Diaminoheptanedioic Acid, 2,6-Diaminoheptanedioic acid

D0112-5G D0112-1G
TCD0112-005GEA 945.33 CAD
TCD0112-005G TCD0112-001G
2,6-Diaminopimelic acid ≥98.0% (by titrimetric analysis)
2,6-Diaminopimelic acid

CAS Number: 583-93-7
MDL Number: MFCD00002637
Molecular Formula: C7H14N2O4
Molecular Weight: 190.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 1,5-Diaminoheptanedioic Acid

Formula: C₇H₁₄N₂O₄
MW: 190.20 g/mol
MDL Number: MFCD00002637
CAS Number: 583-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7da0e57392ef3eba30ada3a8b96441f4e3ddc080&vid=7da0e57392ef3eba30ada3a8b96441f4e3ddc080&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR