2,4-Diaminodiphenylamine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D0087-5G
D0087-25G
TCD0087-5GEA
255.96
CAD
TCD0087-5G
TCD0087-025G
2,4-Diaminodiphenylamine ≥98.0% (by titrimetric analysis)
2,4-Diaminodiphenylamine
CAS Number: 136-17-4
MDL Number: MFCD00025285
Molecular Formula: C12H13N3
Molecular Weight: 199.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 125
Formula:
C₁₂H₁₃N₃ Melting Pt: 125 °C |
MDL Number:
MFCD00025285 CAS Number: 136-17-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...