N,N'-Diacetyl-1,4-phenylenediamine ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

D0068-25G D0068-5G
TCD0068-025GEA 271.76 CAD
TCD0068-025G TCD0068-5G
N,N'-Diacetyl-1,4-phenylenediamine ≥98.0% (by GC, total nitrogen)
N,N'-Diacetyl-1,4-phenylenediamine

CAS Number: 140-50-1
MDL Number: MFCD00026142
Molecular Formula: C10H12N2O2
Molecular Weight: 192.22
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal


Other Product Names / Synonyms: N,N'-1,4-Phenylenebisacetamide

Formula: C₁₀H₁₂N₂O₂
MDL Number: MFCD00026142
CAS Number: 140-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ff88565c3e6a08700721af8ade8094cf8a81a8f&vid=7ff88565c3e6a08700721af8ade8094cf8a81a8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR