D-(+)-Fucose ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
alpha-D-Fucose
D0049-1G
D0049-5G
TCD0049-001GEA
218.66
CAD
TCD0049-001G
TCD0049-005G
D-(+)-Fucose ≥98.0% (by HPLC)
D(+)-Fucose
CAS Number: 3615-37-0
MDL Number: MFCD00135603
Molecular Formula: C6H12O5
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 137
Specific rotation [a]20/D: 75 deg (C=1, H2O)
Product Categories: Bioscience, Fucose, Deoxysugars
Other Product Names / Synonyms: 6-Deoxy-D-galactose, D-(+)-Rhodeose
Formula:
C₆H₁₂O₅ Melting Pt: 137 °C |
MDL Number:
MFCD00135603 CAS Number: 3615-37-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...