1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1-(2-Chloroethyl)hexamethyleneimine Hydrochloride, 1-(2-Chloroethyl)azepane hydrochloride

C2379-25G C2379-5G
TCC2379-25GEA 93.22 CAD
TCC2379-25G TCC2379-5G
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride ≥98.0%
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride

CAS Number: 26487-67-2
MDL Number: MFCD00012842
Molecular Formula: C8H16ClN
Molecular Weight: 198.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 208


Other Product Names / Synonyms: 1-(2-Chloroethyl)hexamethyleneimine Hydrochloride

Formula: C₈H₁₆ClN
MW: 198.13 g/mol
Melting Pt: 208 °C
MDL Number: MFCD00012842
CAS Number: 26487-67-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cf7bead6976b75953f6eb6d54f833a4410e1238&vid=7cf7bead6976b75953f6eb6d54f833a4410e1238&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR