3-Chloro-6-methoxypyridazine ≥97.0%

Supplier: TCI America

C2376-5G C2376-25G
TCC2376-5GEA 120.08 CAD
TCC2376-5G TCC2376-25G
3-Chloro-6-methoxypyridazine ≥97.0%
3-Chloro-6-methoxypyridazine

CAS Number: 1722-10-7
MDL Number: MFCD00006467
Molecular Formula: C5H5ClN2O
Molecular Weight: 144.56
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 175
Melting point (°C): 91

Formula: C₅H₅ClN₂O
Boiling Pt: 175 °C
Melting Pt: 91 °C
MDL Number: MFCD00006467
CAS Number: 1722-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6d2bc83721fa6e251709228b66b27d9b742b337&vid=e6d2bc83721fa6e251709228b66b27d9b742b337&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR