Cyclopropanecarboxamidine hydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

C2344-5G C2344-1G
TCC2344-5GEA 82.16 CAD
TCC2344-5G TCC2344-1G
Cyclopropanecarboxamidine hydrochloride ≥98.0% (by total nitrogen basis)
Cyclopropanecarboxamidine hydrochloride

CAS Number: 57297-29-7
MDL Number: MFCD00053010
Molecular Formula: C4H8N2
Molecular Weight: 120.58
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 127


Other Product Names / Synonyms: Cyclopropanecarboximidamide Hydrochloride

Formula: C₄H₈N₂
Melting Pt: 127 °C
MDL Number: MFCD00053010
CAS Number: 57297-29-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c99133f7eaabea9203dcbf346a6e8aca6d245e33&vid=c99133f7eaabea9203dcbf346a6e8aca6d245e33&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR