3-Chloropicolinic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Chloropyridine-2-carboxylic acid

C2342-5G C2342-1G
TCC2342-5GEA 221.52 CAD
TCC2342-5G TCC2342-1G
3-Chloropicolinic acid ≥98.0%
3-Chloropicolinic acid

CAS Number: 57266-69-0
MDL Number: MFCD01646325
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 154


Other Product Names / Synonyms: 3-Chloropicolinic Acid

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: 154 °C
MDL Number: MFCD01646325
CAS Number: 57266-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c04d6e4854d2e40866dcd086569cdbfb079427fe&vid=c04d6e4854d2e40866dcd086569cdbfb079427fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR