2-Chloro-5-iodopyridine ≥98.0%

Supplier: TCI America

C2341-5G C2341-25G
TCC2341-5GEA 120.71 CAD
TCC2341-5G TCC2341-25G
2-Chloro-5-iodopyridine ≥98.0%
2-Chloro-5-iodopyridine

CAS Number: 69045-79-0
MDL Number: MFCD01863635
Molecular Formula: C5H3ClIN
Molecular Weight: 239.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 99

Formula: C₅H₃ClIN
MW: 239.44
Melting Pt: 99 °C
MDL Number: MFCD01863635
CAS Number: 69045-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=013cd4a979a8c0cf86402a2581d25952eeabddd4&vid=013cd4a979a8c0cf86402a2581d25952eeabddd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR