3-Chloro-2-hydroxypyridine ≥98.0%

Supplier: TCI America

C2340-1G C2340-5G
TCC2340-1GEA 71.1 CAD
TCC2340-1G TCC2340-5G
3-Chloro-2-hydroxypyridine ≥98.0%
3-Chloro-2-hydroxypyridine

CAS Number: 13466-35-8
MDL Number: MFCD01646109
Molecular Formula: C5H4ClNO
Molecular Weight: 129.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 177


Other Product Names / Synonyms: 3-Chloro-2(1H)-pyridinone, 3-Chloro-2-pyridinol, 3-Chloro-2-pyridone

Formula: C₅H₄ClNO
Melting Pt: 177 °C
MDL Number: MFCD01646109
CAS Number: 13466-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a981d4ea700eb2d5935cc7bb4f76506868fb0cd&vid=2a981d4ea700eb2d5935cc7bb4f76506868fb0cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR