3-Chloro-4-iodobenzotrifluoride ≥97.0% stabilized

Supplier: TCI America

C2336-1G C2336-5G
TCC2336-1GEA 41.08 CAD
TCC2336-1G TCC2336-5G
3-Chloro-4-iodobenzotrifluoride ≥97.0% stabilized
3-Chloro-4-iodobenzotrifluoride

(stabilized with Copper chip)
CAS Number: 141738-80-9
MDL Number: MFCD00042562
Molecular Formula: C7H3ClF3I
Molecular Weight: 306.45
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 110
Specific Gravity (20/20): 1.98


Other Product Names / Synonyms: 2-Chloro-1-iodo-4-(trifluoromethyl)benzene, 3-Chloro-4-iodo-alpha,alpha,alpha-trifluorotoluene

Formula: C₇H₃ClF₃I
MW: 306.45 g/mol
Boiling Pt: 60 °C
Density: 1.98 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00042562
CAS Number: 141738-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69d9628b0ba6ad7bd067efb0cb17fcfd49d9efbc&vid=69d9628b0ba6ad7bd067efb0cb17fcfd49d9efbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR