2-Chloro-3-nitrobenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: Mesalazine EP Impurity Q

C2333-5G C2333-25G
TCC2333-5GEA 491.38 CAD
TCC2333-5G TCC2333-25G
2-Chloro-3-nitrobenzoic acid ≥98.0%
2-Chloro-3-nitrobenzoic acid

CAS Number: 3970-35-2
MDL Number: MFCD00007069
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 185

Formula: C₇H₄ClNO₄
Melting Pt: 185 °C
MDL Number: MFCD00007069
CAS Number: 3970-35-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e951d8ffdab44df7671cf870d2f01127e748949&vid=e9f2949f479f4d55a017b2a16b13f27872222576&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR