3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)
C2328-25G
C2328-5G
TCC2328-25GEA
294.2
CAD
TCC2328-25G
TCC2328-5G
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one ≥98.0%
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
CAS Number: 69225-59-8
MDL Number: MFCD00006652
Molecular Formula: C11H18O3
Molecular Weight: 198.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 113
Other Product Names / Synonyms: 3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Formula:
C₁₁H₁₈O₃ MW: 198.26 g/mol Melting Pt: 51 °C Flash Pt: 113 °C |
MDL Number:
MFCD00006652 CAS Number: 69225-59-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...