5-Chloro-1-methylimidazole ≥98.0%

Supplier: TCI America

C2317-5G C2317-25G
TCC2317-5GEA 97.49 CAD
TCC2317-5G TCC2317-25G
5-Chloro-1-methylimidazole ≥98.0%
5-Chloro-1-methylimidazole

CAS Number: 872-49-1
MDL Number: MFCD00014505
Molecular Formula: C4H5ClN2
Molecular Weight: 116.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 96
Specific Gravity (20/20): 1.26

Formula: C₄H₅ClN₂
Boiling Pt: 85 °C
Density: 1.26 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00014505
CAS Number: 872-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4903b7d877e247129fa5d85a83fa224afbcea77c&vid=4903b7d877e247129fa5d85a83fa224afbcea77c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR