6-Chloro-7-deazapurine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine
C2306-5G
C2306-1G
TCC2306-5GEA
588.39
CAD
TCC2306-5G
TCC2306-1G
6-Chloro-7-deazapurine ≥98.0% (by GC, titration analysis)
6-Chloro-7-deazapurine
CAS Number: 3680-69-1
MDL Number: MFCD01686865
Molecular Formula: C6H4ClN3
Molecular Weight: 153.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 188
Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 4-Chloro-7H-pyrrolo[2,3-d]pyrimidine
Formula:
C₆H₄ClN₃ MW: 153.57 g/mol Melting Pt: 188 °C |
MDL Number:
MFCD01686865 CAS Number: 3680-69-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...