4-Chloro-2-iodoanisole ≥98.0%

Supplier: TCI America

C2298-25G C2298-5G
TCC2298-25GEA 399.74 CAD
TCC2298-25G TCC2298-5G
4-Chloro-2-iodoanisole ≥98.0%
4-Chloro-2-iodoanisole

CAS Number: 52807-27-9
MDL Number: MFCD00052858
Molecular Formula: C7H6ClIO
Molecular Weight: 268.48
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 47


Other Product Names / Synonyms: 4-Chloro-2-iodo-1-methoxybenzene

Formula: C₇H₆ClIO
Boiling Pt: 135 °C
Melting Pt: 47 °C
MDL Number: MFCD00052858
CAS Number: 52807-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64117cdf0a67610b458bf1bdfbad8451d6128727&vid=64117cdf0a67610b458bf1bdfbad8451d6128727&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR