5-Chloro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C2292-25G
C2292-5G
TCC2292-25GEA
363.4
CAD
TCC2292-25G
TCC2292-5G
5-Chloro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
5-Chloro-2-methoxyphenylboronic acid
CAS Number: 89694-48-4
MDL Number: MFCD01318966
Molecular Formula: C7H8BClO3
Molecular Weight: 186.40
Form: Crystal
Color: White
Melting point (°C): 145
Other Product Names / Synonyms: 5-Chloro-2-methoxybenzeneboronic Acid
Formula:
C₇H₈BClO₃ Melting Pt: 145 °C |
MDL Number:
MFCD01318966 CAS Number: 89694-48-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...