2-Chloroquinoline-3-carbaldehyde ≥98.0%
Supplier: TCI America
C2289-1G
C2289-5G
TCC2289-1GEA
33.18
CAD
TCC2289-1G
TCC2289-5G
2-Chloroquinoline-3-carbaldehyde ≥98.0%
2-Chloroquinoline-3-carbaldehyde
CAS Number: 73568-25-9
MDL Number: MFCD00130079
Molecular Formula: C10H6ClNO
Molecular Weight: 191.61
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 150
Formula:
C₁₀H₆ClNO Melting Pt: 150 °C |
MDL Number:
MFCD00130079 CAS Number: 73568-25-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...