2-Chloro-5-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C2288-1G
C2288-5G
TCC2288-1GEA
69.52
CAD
TCC2288-1G
TCC2288-5G
2-Chloro-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Chloro-5-pyridineboronic acid
CAS Number: 444120-91-6
MDL Number: MFCD03094998
Molecular Formula: C5H5BClNO2
Molecular Weight: 157.36
Form: Crystal
Color: White
Melting point (°C): 167
Other Product Names / Synonyms: 6-Chloro-3-pyridylboronic Acid
Formula:
C₅H₅BClNO₂ Melting Pt: 167 °C |
MDL Number:
MFCD03094998 CAS Number: 444120-91-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...