2-Chloro-4-nitropyridine ≥98.0%

Supplier: TCI America

C2283-1G C2283-5G C2283-25G
TCC2283-1GEA 60.04 CAD
TCC2283-1G TCC2283-5G TCC2283-25G
2-Chloro-4-nitropyridine ≥98.0%
2-Chloro-4-nitropyridine

CAS Number: 23056-36-2
MDL Number: MFCD00661454
Molecular Formula: C5H3ClN2O2
Molecular Weight: 158.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 106

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 54 °C
Flash Pt: 106 °C
MDL Number: MFCD00661454
CAS Number: 23056-36-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=890be5df1425423f2f0644ef14b170855128cfb3&vid=890be5df1425423f2f0644ef14b170855128cfb3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR