4-(Chloromethyl)-3,5-dimethylisoxazole ≥95.0%
Supplier: TCI America
C2282-5G
C2282-1G
TCC2282-5GEA
290.88
CAD
TCC2282-5G
TCC2282-1G
4-(Chloromethyl)-3,5-dimethylisoxazole ≥95.0%
4-(Chloromethyl)-3,5-dimethylisoxazole
CAS Number: 19788-37-5
MDL Number: MFCD00003154
Molecular Formula: C6H8ClNO
Molecular Weight: 145.59
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 96
Specific Gravity (20/20): 1.17
Storage Temperature: 0-10°C
Formula:
C₆H₈ClNO Boiling Pt: 88 °C Density: 1.17 (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00003154 CAS Number: 19788-37-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...