2-Chloro-N-methoxy-N-methylacetamide ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-Chloro-N-methylacetohydroxamate

C2272-5G C2272-25G
TCC2272-5GEA 63.2 CAD
TCC2272-5G TCC2272-25G
2-Chloro-N-methoxy-N-methylacetamide ≥98.0%
2-Chloro-N-methoxy-N-methylacetamide

CAS Number: 67442-07-3
MDL Number: MFCD00134232
Molecular Formula: C4H8ClNO2
Molecular Weight: 137.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 95
Melting point (°C): 39
Flash Point (°C): 104
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Methyl 2-Chloro-N-methylacetohydroxamate

Formula: C₄H₈ClNO₂
MW: 137.56 g/mol
Boiling Pt: 95 °C
Melting Pt: 39 °C
Flash Pt: 104 °C
MDL Number: MFCD00134232
CAS Number: 67442-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45fd7835d60be4501686f61c0b5cd7d380865726&vid=45fd7835d60be4501686f61c0b5cd7d380865726&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR