2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride

C2269-5G C2269-25G
TCC2269-5GEA 50.56 CAD
TCC2269-5G TCC2269-25G
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride ≥98.0%
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride

CAS Number: 86604-75-3
MDL Number: MFCD00277472
Molecular Formula: C9H12ClNO
Molecular Weight: 222.11
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 131

Formula: C₉H₁₂ClNO
Melting Pt: 131 °C
MDL Number: MFCD00277472
CAS Number: 86604-75-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6910e30ce8bed682da6447c891be46533700a9fc&vid=6910e30ce8bed682da6447c891be46533700a9fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR