Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)
Supplier: TCI America
C2251-1G
C2251-250MG
TCC2251-1GEA
191.18
CAD
TCC2251-1G
TCC2251-250MG
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)
CAS Number: 25360-32-1
MDL Number: MFCD00015870
Molecular Formula: C55H47OP3Ru
Molecular Weight: 917.97
Form: Crystal
Color: White
Melting point (°C): 163
Formula:
C₅₅H₄₇OP₃Ru Melting Pt: 163 °C |
MDL Number:
MFCD00015870 CAS Number: 25360-32-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...