2-Cyanophenylacetonitrile ≥98.0%

Supplier: TCI America

C2247-25G C2247-5G
TCC2247-25GEA 338.91 CAD
TCC2247-25G TCC2247-5G
2-Cyanophenylacetonitrile ≥98.0%
2-Cyanophenylacetonitrile

CAS Number: 3759-28-2
MDL Number: MFCD00001895
Molecular Formula: C9H6N2
Molecular Weight: 142.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 80


Other Product Names / Synonyms: alpha-Cyano-o-tolunitrile, Homophthalonitrile

Formula: C₉H₆N₂
Melting Pt: 80 °C
MDL Number: MFCD00001895
CAS Number: 3759-28-2
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77934a68b2701c7de3c90e3dbcccce22ced260d5&vid=77934a68b2701c7de3c90e3dbcccce22ced260d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR