2-Chloro-3-iodopyridine ≥98.0%
Supplier: TCI America
C2193-5G
C2193-1G
TCC2193-5GEA
123.24
CAD
TCC2193-5G
TCC2193-1G
2-Chloro-3-iodopyridine ≥98.0%
2-Chloro-3-iodopyridine
CAS Number: 78607-36-0
MDL Number: MFCD00661298
Molecular Formula: C5H3ClIN
Molecular Weight: 239.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 95
Formula:
C₅H₃ClIN MW: 239.44 g/mol Melting Pt: 95 °C |
MDL Number:
MFCD00661298 CAS Number: 78607-36-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...