3-Chloro-4-hydroxybenzaldehyde ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2183-5G C2183-25G
TCC2183-5GEA 529.3 CAD
TCC2183-5G TCC2183-25G
3-Chloro-4-hydroxybenzaldehyde ≥97.0% (by GC, titration analysis)
3-Chloro-4-hydroxybenzaldehyde

CAS Number: 2420-16-8
MDL Number: MFCD00016981
Molecular Formula: C7H5ClO2
Molecular Weight: 156.57
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 133

Formula: C₇H₅ClO₂
Boiling Pt: 150 °C
Melting Pt: 133 °C
MDL Number: MFCD00016981
CAS Number: 2420-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7360c19003c5d4a3e1d0f8db0039117d5ad8a486&vid=7360c19003c5d4a3e1d0f8db0039117d5ad8a486&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR