3-Chloro-4-iodoaniline ≥95.0%

Supplier: TCI America

C2167-25G C2167-5G
TCC2167-25GEA 200.66 CAD
TCC2167-25G TCC2167-5G
3-Chloro-4-iodoaniline ≥95.0%
3-Chloro-4-iodoaniline

CAS Number: 135050-44-1
MDL Number: MFCD00051845
Molecular Formula: C6H5ClIN
Molecular Weight: 253.47
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 70

Formula: C₆H₅ClIN
MW: 253.47 g/mol
Boiling Pt: 289
Melting Pt: 70 °C
MDL Number: MFCD00051845
CAS Number: 135050-44-1
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8474fc39633ca1ec055eb9a432c5e4c8b3a2ec49&vid=8474fc39633ca1ec055eb9a432c5e4c8b3a2ec49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR