4-Chloro-m-xylene ≥98.0%
Supplier: TCI America
C2165-25G
C2165-5G
TCC2165-25GEA
262.28
CAD
TCC2165-25G
TCC2165-5G
4-Chloro-m-xylene ≥98.0%
4-Chloro-m-xylene
CAS Number: 95-66-9
MDL Number: MFCD00060644
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Melting point (°C): -32
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1-Chloro-2,4-dimethylbenzene
Formula:
C₈H₉Cl Boiling Pt: 188 °C Melting Pt: –32 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD00060644 CAS Number: 95-66-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...