Carbon nanotube multi-walled 20-40nm (diam.), 5-15µm (length)

Supplier: TCI America

C2154-1G C2154-5G
TCC2154-1GEA 121.66 CAD
TCC2154-1G TCC2154-5G
Carbon nanotube multi-walled 20-40nm (diam.), 5-15µm (length)
Carbon nanotube

CAS Number: 308068-56-6
Form: Powder
Color: Black


Other Product Names / Synonyms: Multi-walled Carbon Nanotube

Formula: C
MDL Number: MFCD00133992
CAS Number: 308068-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR