Carbon nanotube multi-walled 10-20nm (diam.), 5-15µm (length)
Supplier: TCI America
C2150-1G
C2150-5G
TCC2150-1GEA
145.68
CAD
TCC2150-1G
TCC2150-5G
Carbon nanotube multi-walled 10-20nm (diam.), 5-15µm (length)
Carbon nanotube
CAS Number: 308068-56-6
Form: Powder
Color: Black
Other Product Names / Synonyms: Multi-walled Carbon Nanotube
Formula:
C |
MDL Number:
MFCD00133992 CAS Number: 308068-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...