N-Carbobenzoxy-D-valine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C2139-25G C2139-5G
TCC2139-25GEA 161.16 CAD
TCC2139-25G TCC2139-5G
N-Carbobenzoxy-D-valine ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-D-valine

CAS Number: 1685-33-2
MDL Number: MFCD00065703
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 61
Specific rotation [a]20/D: 4 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-valine, Z-D-Val-OH

Formula: C₁₃H₁₇NO₄
Melting Pt: 61 °C
MDL Number: MFCD00065703
CAS Number: 1685-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c94d1e58b6f40c82831df20969e29825756d8109&vid=c94d1e58b6f40c82831df20969e29825756d8109&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR