N-Carbobenzoxy-D-threonine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C2138-1G
C2138-5G
TCC2138-1GEA
57.83
CAD
TCC2138-1G
TCC2138-5G
N-Carbobenzoxy-D-threonine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-D-threonine
CAS Number: 80384-27-6
MDL Number: MFCD00037808
Molecular Formula: C12H15NO5
Molecular Weight: 253.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 104
Specific rotation [a]20/D: 5 deg (C=2, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-threonine, Z-D-Thr-OH
Formula:
C₁₂H₁₅NO₅ Melting Pt: 104 °C |
MDL Number:
MFCD00037808 CAS Number: 80384-27-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...