Cbz-D-Ser-OH ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (R)-2-(((Benzyloxy)carbonyl)amino)-3-hydroxypropanoic acid, N-Cbz-D-serine, Z-D-Ser-OH

C2137-1G C2137-5G
TCC2137-1GEA 33.18 CAD
TCC2137-1G TCC2137-5G
Cbz-D-Ser-OH ≥98.0% (by titrimetric analysis)
Cbz-D-Ser-OH

CAS Number: 6081-61-4
MDL Number: MFCD00063144
Molecular Formula: C11H13NO5
Molecular Weight: 239.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 118
Specific rotation [a]20/D: -6 deg (C=6, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-serine, Z-D-Ser-OH

Formula: C₁₁H₁₃NO₅
MW: 239.2 g/mol
Melting Pt: 118 °C
MDL Number: MFCD00063144
CAS Number: 6081-61-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4f81f859da41cae8bf56503a57d8db543bf2c9c&vid=e4f81f859da41cae8bf56503a57d8db543bf2c9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR