N(α)-Cbz-D-histidine ≥98.0%
Supplier: TCI America
Synonyms:
Z-D-His-OH, N(α)-Benzyloxycarbonyl-D-histidine
C2133-1G
C2133-5G
TCC2133-1GEA
93.22
CAD
TCC2133-1G
TCC2133-5G
N(α)-Cbz-D-histidine ≥98.0%
N(α)-Cbz-D-histidine
CAS Number: 67424-93-5
MDL Number: MFCD00067702
Molecular Formula: C14H15N3O4
Molecular Weight: 289.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 163
Specific rotation [a]20/D: 24 deg (C=6, 6mol/L HCl)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-D-histidine, Z-D-His-OH
Formula:
C₁₄H₁₅N₃O₄ MW: 289.3 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD00067702 CAS Number: 67424-93-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...