2-Chloro-6-fluoroaniline ≥98.0%

Supplier: TCI America

C2126-1G C2126-5G
TCC2126-1GEA 46.29 CAD
TCC2126-1G TCC2126-5G
2-Chloro-6-fluoroaniline ≥98.0%
2-Chloro-6-fluoroaniline

CAS Number: 363-51-9
MDL Number: MFCD00040309
Molecular Formula: C6H5ClFN
Molecular Weight: 145.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 91
Melting point (°C): 32

Formula: C₆H₅ClFN
Boiling Pt: 91 °C
Melting Pt: 32 °C
MDL Number: MFCD00040309
CAS Number: 363-51-9
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR