3-Chloro-2-fluorobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2117-25G C2117-5G
TCC2117-25GEA 132.72 CAD
TCC2117-25G TCC2117-5G
3-Chloro-2-fluorobenzoic acid ≥97.0% (by GC, titration analysis)
3-Chloro-2-fluorobenzoic acid

CAS Number: 161957-55-7
MDL Number: MFCD00042506
Molecular Formula: C7H4ClFO2
Molecular Weight: 174.56
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 177

Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00042506
CAS Number: 161957-55-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d41a6aaab24f1b4cf87894e4f6c814ed6d6129ad&vid=d41a6aaab24f1b4cf87894e4f6c814ed6d6129ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR