5-Chloro-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2094-5G C2094-25G
TCC2094-5GEA 97.49 CAD
TCC2094-5G TCC2094-25G
5-Chloro-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
5-Chloro-2-fluorobenzoic acid

CAS Number: 394-30-9
MDL Number: MFCD00665762
Molecular Formula: C7H4ClFO2
Molecular Weight: 174.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 151

Formula: C₇H₄ClFO₂
Melting Pt: 151 °C
MDL Number: MFCD00665762
CAS Number: 394-30-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68e5a75e901263bdd28df3938cd3781d33632667&vid=68e5a75e901263bdd28df3938cd3781d33632667&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR