2-Chloro-5-fluorobenzoic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

C2088-25G C2088-5G
TCC2088-25GEA 200.66 CAD
TCC2088-25G TCC2088-5G
2-Chloro-5-fluorobenzoic acid ≥95.0% (by titrimetric analysis)
2-Chloro-5-fluorobenzoic acid

CAS Number: 2252-50-8
MDL Number: MFCD00156967
Molecular Formula: C7H4ClFO2
Molecular Weight: 174.56
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 150

Formula: C₇H₄ClFO₂
MW: 174.56
Melting Pt: 150 °C
MDL Number: MFCD00156967
CAS Number: 2252-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5ff7d3c4d06cce9127a332cfa7043fecb3c4084&vid=b5ff7d3c4d06cce9127a332cfa7043fecb3c4084&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR