4-Chloro-2-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2076-25G C2076-5G
TCC2076-25GEA 222.78 CAD
TCC2076-25G TCC2076-5G
4-Chloro-2-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Chloro-2-methoxybenzoic acid

CAS Number: 57479-70-6
MDL Number: MFCD00002532
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: 4-Chloro-o-anisic Acid

Formula: C₈H₇ClO₃
Melting Pt: 148 °C
MDL Number: MFCD00002532
CAS Number: 57479-70-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1925b6e0b40540d22570d253c7eadd3eeb28c8f1&vid=1925b6e0b40540d22570d253c7eadd3eeb28c8f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR