5-Chloro-2-thiopheneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(5-Chlorothiophen-2-yl)boronic acid, 5-Chloro-2-thienylboronic acid, 5-Chlorothiophene-2-boronic acid
C2066-1G
C2066-5G
TCC2066-1GEA
127.98
CAD
TCC2066-1G
TCC2066-5G
5-Chloro-2-thiopheneboronic acid (contains varying amounts of Anhydride)
5-Chloro-2-thiopheneboronic acid
CAS Number: 162607-18-3
MDL Number: MFCD00236030
Molecular Formula: C4H4BClO2S
Molecular Weight: 162.39
Form: Crystal
Color: White
Melting point (°C): 140
Formula:
C₄H₄BClO₂S Melting Pt: 140 °C |
MDL Number:
MFCD00236030 CAS Number: 162607-18-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...