Cerium trichloride, anhydrous ≥96.0%
Supplier: TCI America
Synonyms:
Cerous chloride, Cerium(III) chloride, Cerium (III) chloride
C2058-25G
C2058-5G
TCC2058-25GEA
188.05
CAD
TCC2058-25G
TCC2058-5G
Cerium trichloride, anhydrous ≥96.0%
Cerium trichloride
CAS Number: 7790-86-5
Molecular Formula: CeCl3
Molecular Weight: 246.47
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 722
Other Product Names / Synonyms: Cerous Chloride
Formula:
CeCl₃ MW: 246.47 g/mol Melting Pt: 848 |
MDL Number:
MFCD00010929 CAS Number: 7790-86-5 Merck Index: 13,02008 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...