3-Chloro-o-toluic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2057-5G C2057-1G
TCC2057-5GEA 160.37 CAD
TCC2057-5G TCC2057-1G
3-Chloro-o-toluic acid ≥97.0% (by GC, titration analysis)
3-Chloro-o-toluic acid

CAS Number: 7499-08-3
MDL Number: MFCD00053312
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 164


Other Product Names / Synonyms: 3-Chloro-o-toluic Acid

Formula: C₈H₇ClO₂
Melting Pt: 164 °C
MDL Number: MFCD00053312
CAS Number: 7499-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cde78df21ab4cbbd65ec9a2560c5a6d4d62c2e80&vid=cde78df21ab4cbbd65ec9a2560c5a6d4d62c2e80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR