6-Chloroanthranilic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C2048-25G
C2048-5G
TCC2048-25GEA
105.86
CAD
TCC2048-25G
TCC2048-5G
6-Chloroanthranilic acid ≥98.0% (by HPLC, titration analysis)
6-Chloroanthranilic acid
CAS Number: 2148-56-3
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 142
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2-Amino-6-chlorobenzoic Acid
Formula:
C₇H₆ClNO₂ Melting Pt: 142 °C |
MDL Number:
MFCD00051530 CAS Number: 2148-56-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...