2-Chloro-5-(trifluoromethyl)benzaldehyde ≥95.0%
Supplier: TCI America
C2047-5G
C2047-25G
TCC2047-5GEA
99.54
CAD
TCC2047-5G
TCC2047-25G
2-Chloro-5-(trifluoromethyl)benzaldehyde ≥95.0%
2-Chloro-5-(trifluoromethyl)benzaldehyde
CAS Number: 82386-89-8
MDL Number: MFCD00134430
Molecular Formula: C8H4ClF3O
Molecular Weight: 208.56
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 89
Specific Gravity (20/20): 1.45
Formula:
C₈H₄ClF₃O Boiling Pt: 97 °C Density: 1.45 (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00134430 CAS Number: 82386-89-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...