3-Carboxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

C2028-1G C2028-5G
TCC2028-1GEA 61.62 CAD
TCC2028-1G TCC2028-5G
3-Carboxyphenylboronic acid (contains varying amounts of Anhydride)
3-Carboxyphenylboronic acid

CAS Number: 25487-66-5
Molecular Formula: C7H7BO4
Molecular Weight: 165.94
Form: Crystal
Color: White
Melting point (°C): 240


Other Product Names / Synonyms: 3-Carboxybenzeneboronic Acid

Formula: C₇H₇BO₄
Melting Pt: 240 °C
MDL Number: MFCD00036833
CAS Number: 25487-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7351e06fba03dca76433a1892fab3452ad1059dd&vid=7351e06fba03dca76433a1892fab3452ad1059dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR